Skip to Content
Merck
All Photos(2)

Documents

06-1358

Sigma-Aldrich

Anti-acetyl (Lys34) phospho (Ser36) Histone H1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H1.4, Histone H1b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, bovine

species reactivity (predicted by homology)

canine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer36), acetylation (Lys34)

Gene Information

bovine ... H1-4(617854)
dog ... H1-4(488269)
human ... H1-4(3008)
rhesus monkey ... H1-4(696872)

General description

H1.4 is one of four variants of histone H1 (H1.2-H1.5) that is found in somatic cells. The H1 histones may function as linker proteins binding DNA via their GH domain, and enabling the wrapping of DNA around the core nucleosomal structure formed from the association of H2A, H2B, H3, and H4. In response to various stimuli, histones undergo a number of post-translational modifications (PTM) including acetylation, phoshorylation, and methylation. Previous studies have suggested that H1.4 may be phosphorylated during mitosis and interphase resulting in increased transcription of target genes by RNA polymerase I and II, and therefore this PTM on H1.4 may have implications for cell growth. H1.4 may also undergo acetylation of its N-terminal lysine residues. Often, it is the pattern of different modifications on several histone residues that alter chromatin and affect gene expression, rather than modification of single residues.

Specificity

Other homologies: Mouse and Rat (90% sequence homology).
This antibody recognizes Histone H1 acetylated at Lys34 and phosphorylated at Ser36.

Immunogen

Epitope: Acetylated Lys34 and phosphorylated Ser36
KLH-conjugated linear peptide corresponding to human Histone H1 acetylated at Lys34 and phosphorylated at Ser36.

Application

Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see Table) were probed with acetyl (Lys34) phospho (Ser36) Histone H1.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-acetyl (Lys34) phospho (Ser36) Histone H1 Antibody (Rabbit Polyclonal Antibody) validated in WB & DB to detect acetyl (Lys34) phospho (Ser36) Histone H1 also known as Histone H1b.

Quality

Evaluated by Western Blot in calf thymus histone extract.

Western Blot Analysis: A 1:37 dilution of this antibody detected Histone H1 on 10 µg of calf thymus histone extract.

Target description

~34 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Calf thymus histone extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service