Skip to Content
Merck

COMH04236A2A

Combi-Blocks, Inc.

3-[(tert-Butoxycarbonyl)amino]bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Synonym(s):

3-(tert-butoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H19O4N1
CAS Number:
Molecular Weight:
253.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H19NO4/c1-13(2,3)18-12(17)14-10-8-5-4-7(6-8)9(10)11(15)16/h4-5,7-10H,6H2,1-3H3,(H,14,17)(H,15,16)

Inchi Key

ZPMJBHUNSHDFPT-UHFFFAOYSA-N

Purity

95%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library