Z514462
Safeskin® purple nitrile gloves
size XS
Synonym(s):
disposable glove
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Recommended Products
material
nitrile
purple
size
XS
sterility
non-sterile
feature
disposable
powder-free
latex free
packaging
pack of 100 ea (boxes of 100)
pk of 100 ea
manufacturer/tradename
Kimberly Clark 55080
availability
not available in EU
L
9 1/2 in.
thickness
4.7 mil , palm
5.9 mil , fingertips
color
purple
Looking for similar products? Visit Product Comparison Guide
General description
These unique purple gloves combine the durability of nitrile with superb comfort and fit. Powder free design reduces the risk of powder-related interference during lab work
Legal Information
Safeskin is a registered trademark of Kimberly-Clark Inc.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service