Skip to Content
Merck
All Photos(1)

Key Documents

W249017

Sigma-Aldrich

Furfuryl acetate

natural, ≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
FEMA Number:
2490
EC Number:
Council of Europe no.:
2065c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.128

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

color

colorless

refractive index

n20/D 1.462 (lit.)

bp

175-177 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana

SMILES string

CC(=O)OCc1ccco1

InChI

1S/C7H8O3/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

CKOYRRWBOKMNRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service