Skip to Content
Merck
All Photos(1)

Documents

NRS3HT8

Norell® Select Series 3 mm high-throughput NMR tube

L 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

material

borosilicate glass (ASTM Type 1 class A)
borosilicate glass tube

feature

disposable

packaging

pkg of 25 ea

manufacturer/tradename

Norell S-3-HT-8

L

8 in.

camber

0.040 mm

diam.

3 mm

tube diam.

3 mm

wall thickness

0.58 mm

Looking for similar products? Visit Product Comparison Guide

General description

The high-throughput NMR tubes are ideal for reducing salt effects when running buffered solutions for bio-samples. They are made from ASTM Type 1 Class A borosilicate glass for reproducibility and durability, and are specially designed for one time use for routine NMR.

Legal Information

Norell is a registered trademark of Norell, Inc.
Select Series is a trademark of Norell, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service