Skip to Content
Merck
All Photos(1)

Key Documents

GF57435318

Stainless Steel - AISI 302 alloy, FeCr18Ni8

foil, 8mm disks, 0.2mm thickness, hard

Synonym(s):

FE200297

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

foil (disks - hard)

manufacturer/tradename

Goodfellow 574-353-18

size × thickness

8 mm × 0.2 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service