Documents
BR144610
BRAND® BISTABIL® ground glass stopper, conical joint
borosilicate glass 3.3, joint: ST/NS 10/19, (hollow, hexagonal grip)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass 3.3
Joint
joint: ST/NS10/19
manufacturer/tradename
BRAND 144610
Related Categories
General description
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences