Skip to Content
Merck
All Photos(1)

Documents

901194

Sigma-Aldrich

(t-Bu)2PMe-Pd-G3

95%

Synonym(s):

Methanesulfonato (di-tert-butyl) methylphosphino (2-amino-1,1-biphenyl-2-yl) palladium(II), Methanesulfonato (di-tert-butyl) methylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34NO3PPdS
CAS Number:
Molecular Weight:
529.97
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

160-173 °C (decomp)

SMILES string

CP(C(C)(C)C)C(C)(C)C.NC1=C(C2=C([Pd]OS(C)(=O)=O)C=CC=C2)C=CC=C1

InChI

1S/C12H10N.C9H21P.CH4O3S.Pd/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-8(2,3)10(7)9(4,5)6;1-5(2,3)4;/h1-6,8-9H,13H2;1-7H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

SSBARXQHJYOPIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Third-generation Buchwald precatalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service