Skip to Content
Merck
All Photos(1)

Key Documents

900132

Sigma-Aldrich

MS Resin®

UV curable 3D printing resin, white

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

size

1 L

color

white

Looking for similar products? Visit Product Comparison Guide

Application

MS Resin® is a new formula that expands what is possible with your printer. The formula is crafted to not be limited to a single printer - many SLA/DLP printers are compatible with MS Resin® including the Form1. The final surface texture has a unique matte finish. The resin is less viscous than most other resins and is easier to clean out from vats and off prints. It′s ideal for anyone who wants to explore superior material options for their UV Curable Printer.

Legal Information

MS Resin is a registered trademark of MadeSolid, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service