Skip to Content
Merck
All Photos(1)

Documents

861502

Sigma-Aldrich

Lucifer Yellow CH dilithium salt

Synonym(s):

6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulfonic acid dilithium salt, Lucifer Yellow CH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Li2N5O9S2
CAS Number:
Molecular Weight:
457.25
Beilstein:
4644601
EC Number:
MDL number:
UNSPSC Code:
12171500

λmax

348 nm
428 nm (2nd)

SMILES string

[Li+].[Li+].NNC(=O)NN1C(=O)c2cc(cc3c(N)c(cc(C1=O)c23)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C13H11N5O9S2.2Li/c14-10-5-1-4(28(22,23)24)2-6-9(5)7(3-8(10)29(25,26)27)12(20)18(11(6)19)17-13(21)16-15;;/h1-3H,14-15H2,(H2,16,17,21)(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2

InChI key

RPKCZJYDUKVMGF-UHFFFAOYSA-L

Application

Highly fluorescent dye for marking nerve cells. The dye spreads rapidly through the injected cell and binds effectively to tissue by a variety of fixatives.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W W Stewart
Nature, 292(5818), 17-21 (1981-07-02)
Lucifer dyes are intensity fluorescent 4-aminonaphthalimides which are readily visible in living cells at concentrations and levels of illumination at which they are nontoxic. Because of their low molecular weight they frequently pass from one cell to another; this widespread

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service