Skip to Content
Merck
All Photos(2)

Documents

807737

Sigma-Aldrich

Photo-reactive Clickable cis-Sterol Probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46N2O3
Molecular Weight:
482.70
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

95%

Quality Level

form

semisolid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

C[C@H](CCC(=O)OCCCCC#C)[C@H]1CCC2C3CC4(N=N4)[C@@H]5C[C@H](O)CC[C@]5(C)C3CC[C@]12C

InChI

1S/C30H46N2O3/c1-5-6-7-8-17-35-27(34)12-9-20(2)23-10-11-24-22-19-30(31-32-30)26-18-21(33)13-15-29(26,4)25(22)14-16-28(23,24)3/h1,20-26,33H,6-19H2,2-4H3/t20-,21-,22?,23-,24?,25?,26-,28-,29-/m1/s1

InChI key

ZJMZIZZWLLQHJO-HFMSFSOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photo-activatable sterol probe for the bioorthogonal tagging and enrichment of sterol-binding biomolecules. This sterol probe contains an intact steroid core structure bearing a diazirine UV-activatable cross-linking group, as well as an alkyne group for click-chemistry mediated adduction of reporter and affinity tags. The probes are cell-permeable, allowing for in situ labeling of biomolecules in live cells, as well as in cell extracts in vitro.

Other Notes

Handle the probes in amber vessels under dim ambient light whenever possible. Aqueous solutions of probes complexed to methyl-β-cyclodextrin (20:1 = m-β-CD:probe molar ratio, 5-20 μM probe) permeate cultured cells in complete serum-containing medium within 30 minutes at 37°C and are cross-linked using near-UV (365 nm) light for 3-10 minutes. Cells can then be harvested and lysed, fractionated or otherwise processed. Click chemistry (Cu(I)-catalyzed 1,3-dipolar cycloaddition of the alkyne with an azide reporter tag such as a fluorophore or biotin, products 760765 or 762024) can be performed in protein samples in PBS for gel-visualization of probe-labeling, MS-based proteomics analysis of probe-mediated affinity enrichments or western blot analysis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan J Hulce et al.
Nature methods, 10(3), 259-264 (2013-02-12)
Cholesterol is an essential structural component of cellular membranes and serves as a precursor for several classes of signaling molecules. Cholesterol exerts its effects and is, itself, regulated in large part by engagement in specific interactions with proteins. The full

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service