Skip to Content
Merck
All Photos(1)

Key Documents

727008

Sigma-Aldrich

(R)-4-Chloromandelic acid

ChiPros®, produced by BASF, 98%

Synonym(s):

4-Chloro-D-mandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO3
CAS Number:
Molecular Weight:
186.59
Beilstein:
2692851
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

produced by BASF

Assay

≥98% (HPLC)
98%

form

solid

optical purity

enantiomeric excess: ≥95%

functional group

carboxylic acid
chloro
hydroxyl

SMILES string

O[C@@H](C(O)=O)C1=CC=C(Cl)C=C1

InChI

1S/C8H7ClO3/c9-6-3-1-5(2-4-6)7(10)8(11)12/h1-4,7,10H,(H,11,12)/t7-/m1/s1

InChI key

BWSFWXSSALIZAU-SSDOTTSWSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service