Skip to Content
Merck
All Photos(1)

Key Documents

724378

Sigma-Aldrich

5-Methoxy-1H-benzo[g]indole

96%

Synonym(s):

5-Methoxy-1H-benz[g]indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO
CAS Number:
Molecular Weight:
197.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

mp

124-127 °C

SMILES string

COc1cc2cc[nH]c2c3ccccc13

InChI

1S/C13H11NO/c1-15-12-8-9-6-7-14-13(9)11-5-3-2-4-10(11)12/h2-8,14H,1H3

InChI key

DGGPMXAQMFWTBH-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service