Skip to Content
Merck
All Photos(1)

Key Documents

68916

Sigma-Aldrich

(R)-(−)-S-Methyl-S-phenylsulfoximine

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NOS
CAS Number:
Molecular Weight:
155.22
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

≥99.0% (GC)

form

solid

optical activity

[α]/D −28.5±3, c = 1 in acetonitrile

SMILES string

C[S@@](=N)(=O)c1ccccc1

InChI

1S/C7H9NOS/c1-10(8,9)7-5-3-2-4-6-7/h2-6,8H,1H3/t10-/m1/s1

InChI key

YFYIDTVGWCYSEO-SNVBAGLBSA-N

Other Notes

A new class of C1-symmetric monosulfoximine ligands for enantioselective hetero Diels-Alder reactions: quinoline-based C1-symmetric sulfoximines lead to cycloadducts with up to 96% ee

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carsten Bolm et al.
Chemical communications (Cambridge, England), (22), 2826-2827 (2003-12-04)
Quinoline-based C1-symmetric sulfoximines have been used as chiral ligands in copper-catalyzed asymmetric hetero Diels-Alder reactions leading to cycloadducts with up to 96% ee.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service