Skip to Content
Merck
All Photos(1)

Key Documents

659401

Sigma-Aldrich

Poly(styrene-ran-ethylene), sulfonated, cross-linkable solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4SO3H)]m(CH2CH2)n
MDL number:
UNSPSC Code:
26111700

composition

styrene, 76 wt. %
sulfonated stryene units, 32-38%
vinyl silane cross-linking agent, <0.5%

concentration

5 wt. % in 1-propanol

refractive index

n20/D 1.384

conductivity

0.1184 S/cm (The ionic conductivity can increase up to 50% when heating from RT to 67°C)

bp

57-60 °C

density

0.85 g/mL at 25 °C

Application

Cross-linkable solid polymeric electrolyte.
Product becomes cross-linked with 800-1,000 mJ/cm2 UV exposure after drying at 110 °C for 10-15 min; or at 140-150 °C for 1 min.

Preparation Note

To cast a film:
1. Concentrate solution to 600 centapoise viscosity (typically 6-9 wt.% polymer).
2. Preheat the casting surface to 35 °C. A clean, flat PTFE-coated glass surface is recommended.
3. Secure a silicone release liner to the casting surface.
4. Place a small drop of the polymer solution on the release liner.
5. Draw the polymer solution down the release liner.
6. Dry the film for 30 min. For controlled evaporation it is recommended to cover surface with aluminum foil that has been perforated with small pinholes.
Treat with 0.5M Sulfuric acid for 20 min to strengthen after cross-linking.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service