562653
Methyl (R)-(+)-2-isocyanato-3-phenylpropionate
97%
About This Item
Assay
97%
optical activity
[α]20/D +70°, c = 1.4% in chloroform
refractive index
n20/D >1.159 (lit.)
bp
256-257 °C/760 mmHg (lit.)
density
1.159 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
COC(=O)[C@@H](Cc1ccccc1)N=C=O
InChI
1S/C11H11NO3/c1-15-11(14)10(12-8-13)7-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3/t10-/m1/s1
InChI key
JOTMMIYKEOOTNZ-SNVBAGLBSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
116.1 °F - closed cup
Flash Point(C)
46.7 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service