467987
trans-3-(3-Thienyl)acrylic acid
97%
Synonym(s):
trans-3-Thiopheneacrylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
97%
mp
152-154 °C (lit.)
SMILES string
OC(=O)\C=C\c1ccsc1
InChI
1S/C7H6O2S/c8-7(9)2-1-6-3-4-10-5-6/h1-5H,(H,8,9)/b2-1+
InChI key
VYRYYUKILKRGDN-OWOJBTEDSA-N
General description
trans-3-(3-Thienyl)acrylic acid (thaH) forms [Mn12] complexes having sulfur-containing carboxylates, [Mn12O12(tha)16(H2O)4]. Magnetic property of these complexes has been investigated.
Application
trans-3-(3-Thienyl)acrylic acid may be used in the preparation of p-coumaroyl-CoA analog.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Single-Molecule Magnets: Magnetic Properties of [Mn 12] Complexes with Sulfer-Containing Carboxylates.
Mol. Cryst. Liq. Cryst., 379(1), 179-184 (2002)
Substrate specificity of chalcone synthase: enzymatic formation of unnatural polyketides from synthetic cinnamoyl-CoA analogues.
Journal of the American Chemical Society, 122(45), 11242-11243 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service