Skip to Content
Merck
All Photos(1)

Key Documents

421375

Sigma-Aldrich

Zonyl® FSA fluorosurfactant

25 wt. % Li carboxylate salt in water: isopropyl alcohol (1:1)

Synonym(s):

Lithium 3-[2-(perfluoroalkyl)ethylthio]propionate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002

concentration

25 wt. % Li carboxylate salt in water: isopropyl alcohol (1:1)

refractive index

n20/D 1.382 (lit.)

surface tension

48 dyn/cm, 0.001 wt. % in H2O

bp

~100 °C (lit.)

solubility

10 wt. % KOH: soluble
organic solvents: insoluble

density

1.03 g/mL at 25 °C (lit.)

InChI

1S/C6H9F3O2S.Li/c7-6(8,9)2-4-12-3-1-5(10)11;/h1-4H2,(H,10,11);/q;+1/p-1

InChI key

WEQTXBLWLWRSCD-UHFFFAOYSA-M

General description

Stable to heat, alkalis and oxidizing agents. Improves wetting and leveling.

Legal Information

DuPont Product
Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service