395110
Triisobutylphosphine
95%
Synonym(s):
Tris(2-methylpropyl)phosphine
About This Item
Recommended Products
Assay
95%
reaction suitability
reagent type: ligand
refractive index
n20/D 1.451 (lit.)
bp
126 °C/50 mmHg (lit.)
density
0.811 g/mL at 25 °C (lit.)
functional group
phosphine
SMILES string
CC(C)CP(CC(C)C)CC(C)C
InChI
1S/C12H27P/c1-10(2)7-13(8-11(3)4)9-12(5)6/h10-12H,7-9H2,1-6H3
InChI key
DAGQYUCAQQEEJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service