377139
1,2,3,4-Tetrahydro-9-acridinecarboxylic acid dihydrate
96%
About This Item
Assay
96%
mp
284-288 °C (lit.)
SMILES string
[H]O[H].[H]O[H].OC(=O)c1c2CCCCc2nc3ccccc13
InChI
1S/C14H13NO2.2H2O/c16-14(17)13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13;;/h1,3,5,7H,2,4,6,8H2,(H,16,17);2*1H2
InChI key
GGRKIMLQOSYIDR-UHFFFAOYSA-N
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service