Skip to Content
Merck
All Photos(1)

Key Documents

307149

Sigma-Aldrich

2,3,6,7-Tetrachloroquinoxaline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2Cl4N2
CAS Number:
Molecular Weight:
267.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

174-176 °C (lit.)

SMILES string

Clc1cc2nc(Cl)c(Cl)nc2cc1Cl

InChI

1S/C8H2Cl4N2/c9-3-1-5-6(2-4(3)10)14-8(12)7(11)13-5/h1-2H

InChI key

GQWLQHHUPKCOJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The standard molar enthalpy of formation for 2,3,6,7-tetrachloroquinoxaline from the standard molar enthalpy of combustion in oxygen was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermochemical study of chloropyrazines and chloroquinoxalines.
The Journal of Chemical Thermodynamics, 36(5), 377-383 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service