Skip to Content
Merck
All Photos(1)

Key Documents

29525

Sigma-Aldrich

(S)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine]

≥97%, optical purity ee: ≥99%

Synonym(s):

(S)-2,2′-Bis[bis(3,5-di-tert-butyl)phosphino]-6,6′-dimethoxy-1,1′-biphenyl, (S)-3,5-t-Bu-MeOBIPHEP, SL-A121-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H96O2P2
Molecular Weight:
1031.46
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

optical purity

ee: ≥99%

SMILES string

COc1cccc(P(c2cc(cc(c2)C(C)(C)C)C(C)(C)C)c3cc(cc(c3)C(C)(C)C)C(C)(C)C)c1-c4c(OC)cccc4P(c5cc(cc(c5)C(C)(C)C)C(C)(C)C)c6cc(cc(c6)C(C)(C)C)C(C)(C)C

InChI

1S/C70H96O2P2/c1-63(2,3)45-33-46(64(4,5)6)38-53(37-45)73(54-39-47(65(7,8)9)34-48(40-54)66(10,11)12)59-31-27-29-57(71-25)61(59)62-58(72-26)30-28-32-60(62)74(55-41-49(67(13,14)15)35-50(42-55)68(16,17)18)56-43-51(69(19,20)21)36-52(44-56)70(22,23)24/h27-44H,1-26H3

InChI key

PBYRAYONARLAQJ-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Application

(S)-3,5-t-Bu-MeOBIPHEP can be used as a ligand in the stereoselective preparation of allenoates via palladium-catalyzed asymmetric carbonylation of propargylic carbonates. It can also be used in the C-C coupling reactions.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A room-temperature catalytic asymmetric synthesis of allenes with ECNU-Phos
Wang Y, et al.
Journal of the American Chemical Society, 135(31), 11517-11520 (2013)
Enantioselective Homogeneous Catalysis and the ?3, 5-Dialkyl Meta-Effect?. MeO- BIPHEP Complexes Related to Heck, Allylic Alkylation, and Hydrogenation Chemistry
Trabesinger G, et al.
Journal of the American Chemical Society, 119(27), 6315-6323 (1997)

Articles

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service