268763
Dodecyltrimethylammonium bromide
99%
Synonym(s):
Lauryltrimethylammonium bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)11N(CH3)3Br
CAS Number:
Molecular Weight:
308.34
Beilstein:
3597463
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
99%
mp
246 °C (dec.) (lit.)
SMILES string
[Br-].CCCCCCCCCCCC[N+](C)(C)C
InChI
1S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1
InChI key
XJWSAJYUBXQQDR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jade E LaDow et al.
European journal of medicinal chemistry, 46(9), 4219-4226 (2011-07-29)
A series of cationic amphiphiles, each with an aromatic core, was prepared and investigated for antimicrobial properties. The synthesized amphiphiles in this study are bicephalic (double-headed) in that they each possess two trimethylammonium head groups and a single linear alkoxy
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service