Skip to Content
Merck
All Photos(1)

Key Documents

238392

Sigma-Aldrich

Sodium deoxycholate monohydrate

97%

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO4 · H2O
CAS Number:
Molecular Weight:
432.57
Beilstein:
3643164
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +48.3°, c = 1 in ethanol

SMILES string

O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3CCC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C

InChI

1S/C24H40O4.Na.H2O/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);;1H2/q;+1;/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;;/m1../s1

InChI key

NDVHNZISGVSFMP-WTCAICSISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Powerful solubilization agent; used in the solubilization of estrogen synthetase from human placental microsomes.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service