All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H26N2O3 · HCl · xH2O
CAS Number:
Molecular Weight:
390.90 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
optical activity
[α]21/D −67°, c = 1 in H2O
mp
288-290 °C (lit.)
InChI
1S/C21H26N2O3.ClH.H2O/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24;;/h2-5,12,15,17-19,22,24H,6-11H2,1H3;1H;1H2/t12-,15-,17-,18-,19-;;/m0../s1
InChI key
XUMWCHYKGZMCSP-QVURBYDESA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service