Skip to Content
Merck
All Photos(1)

Documents

214523

Sigma-Aldrich

Acid Red 8

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2Na2O7S2
CAS Number:
Molecular Weight:
480.42
Colour Index Number:
14900
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 40%

λmax

508 nm
532 nm (2nd)

SMILES string

[Na+].[Na+].Cc1ccc(\N=N\c2c(O)c3ccc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)c(C)c1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service