Skip to Content
Merck
All Photos(1)

Key Documents

16297

Sigma-Aldrich

2-(4-Boc-piperazino)-2-(4-methoxyphenyl)acetic acid

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
9718382
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (HPLC)

form

solid

application(s)

peptide synthesis

SMILES string

COc1ccc(cc1)C(N2CCN(CC2)C(=O)OC(C)(C)C)C(O)=O

InChI

1S/C18H26N2O5/c1-18(2,3)25-17(23)20-11-9-19(10-12-20)15(16(21)22)13-5-7-14(24-4)8-6-13/h5-8,15H,9-12H2,1-4H3,(H,21,22)

InChI key

DCFDOKBNIXUWKP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service