Skip to Content
Merck
All Photos(1)

Key Documents

10878

Sigma-Aldrich

1,3-Dichloroacetone solution

produced by Wacker Chemie AG, Burghausen, Germany, 50% in acetone (density determination)

Synonym(s):

1,3-DCA, 1,3-Dichloro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COCH2Cl
CAS Number:
Molecular Weight:
126.97
Beilstein:
605456
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

produced by Wacker Chemie AG, Burghausen, Germany

concentration

50% in acetone (density determination)

bp

73 °C/733.4 mmHg (lit.)

density

1.025 g/mL at 20 °C (lit.)

SMILES string

ClCC(=O)CCl

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service