Merck
  • PRDM14, a putative histone methyl-transferase, interacts with and decreases the stability and activity of the HOXA1 transcription factor.
PRDM14, a putative histone methyl-transferase, interacts with and decreases the stability and activity of the HOXA1 transcription factor.
Biochimica et biophysica acta (2018-02-23)
Amandine Draime, Laure Bridoux, Magali Belpaire, Tamara Pringels, Janne Tys, René Rezsohazy
ABSTRACT
Understanding how the activity of transcription factors like HOX proteins is regulated remains a widely open question. In a recent screen for proteins interacting with HOXA1, we identified a PRDM protein family member, PRDM14, which is known to be transiently co-expressed with HOXA1 in epiblast cells before their specification towards somatic versus germ cell fate. Here, we confirm PRDM14 is an interactor of HOXA1 and we identify the homeodomain of HOXA1 as well as the PR domain and Zinc fingers of PRDM14 to be required for the interaction. An 11-His repeat of HOXA1 previously highlighted to contribute to HOXA1-mediated protein-protein interactions is also involved. At a functional level, we provide evidence that HOXA1 displays an unexpectedly long half-life and demonstrate that PRDM14 can reduce the stability and affect the transcriptional activity of HOXA1.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt >300,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Glutathione Sepharose 4B, Cytiva 17-0756-01, pack of 10 mL
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)