Skip to Content
Merck
All Photos(1)

Key Documents

PHR9304

Supelco

4-(4-Methylphenyl)-4-oxobutyric acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3-p-Toluoylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH2CH2CO2H
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

Not traceable to any primary pharmacopeia standard

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

127-130 °C (lit.)

application(s)

pharmaceutical

storage temp.

15-25°C

SMILES string

Cc1ccc(cc1)C(=O)CCC(O)=O

InChI

1S/C11H12O3/c1-8-2-4-9(5-3-8)10(12)6-7-11(13)14/h2-5H,6-7H2,1H3,(H,13,14)

InChI key

OEEUWZITKKSXAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service