27083-U
Vials, screw top, amber glass (vial only)
volume 2 mL, amber glass vial, thread for 8-425, pkg of 100 ea
Synonym(s):
Amber Glass Storage Vials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
amber glass vial
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mm
volume
2 mL
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
General description
Vials (volume 2 mL, amber glass vial, thread 8-425) are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Amber glass protects sensitive samples from exposure to UV light.
Application
Vials, screw top, amber glass (vial only) has been used in loading samples such as phospholipids oxidized in vitro or extracted from LDL or cells, analyzed by electrospray ionization-mass spectrometry (ESI-MS) and liquid chromatography-tandem quadrupole-linear ion trap (LC-QqLIT-MS).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Identification of oxidized phospholipids by electrospray ionization mass spectrometry and LC?MS using a QQLIT instrument.
Free Radical Biology & Medicine, 51(12), 2133-2149 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service