Skip to Content
Merck
All Photos(1)

Documents

13359-U

Supelco

Supelpak-2 polymeric adsorbent

20-60 mesh, pk of 120g, for Japan only

Synonym(s):

XAD®-2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151817

product name

Supelpak-2, pk of 120 g, for Japan only

Quality Level

description

purified XAD-2

packaging

pk of 120 g

parameter

200 °C max. temp.

matrix

polymeric styrene-divinylbenzene

pore size

~0.65 mL/g pore volume
90 Å mean pore size

density

1.02 g/mL at 25 °C (true wet)(lit.)
1.07 g/mL at 25 °C (skeletal)(lit.)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Supelpak-2 is purified Amberlite XAD-2® that has been cleaned to meet and exceed US EPA-recommended criteria for purity, as outlined in Level I Environmental Assessment Procedures Manual. It is the best resin to use for standard air sampling methods requiring resin tested for background TCO (total chromatographic organics) level. Packaged in glass containers.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service