Z331694
BRAND® macro pipette controller
magenta
Synonym(s):
brand macro pipette, brand pipette
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
magenta
plastic (autoclavable)
silicone (autoclavable)
feature
autoclavable
manufacturer/tradename
BRAND 26203
volume
0.1-200 mL
General description
One single macro pipette controller covers the entire range of bulb and graduated pipettes from 0.1 to 200 mL. The conical silicone adapter offers a secure fit for the different diameters. The unit is fully autoclavable at 121°C (2 bar) according to DIN EN 285.
Supplied with a spare hydrophobic membrane filter which protects the system from liquid penetration.
Supplied with a spare hydrophobic membrane filter which protects the system from liquid penetration.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)