Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100527

Sigma-Aldrich

Anti-DAAM1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Dishevelled associated activator of morphogenesis 1, Anti-FLJ41657, Anti-KIAA0666

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

123 kDa

species reactivity

mouse, horse, guinea pig, bovine, rabbit, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAAM1(23002)

Immunogen

Synthetic peptide directed towards the middle region of human DAAM1

Biochem/physiol Actions

Functions of the cell cortex, including motility, adhesion, and cytokinesis, are mediated by the reorganization of the actin cytoskeleton and recent evidence suggests a role for the Formin homology (FH) proteins in these processes. The protein encoded by this gene contains FH domains and belongs to a novel FH protein subfamily implicated in cell polarity. Wnt/Fz signaling activates the small GTPase Rho, a key regulator of cytoskeleton architecture, to control cell polarity and movement during development. Activation requires Dvl-Rho complex formation, an assembly mediated by this gene product, which is thought to function as a scaffolding protein.Functions of the cell cortex, including motility, adhesion, and cytokinesis, are mediated by the reorganization of the actin cytoskeleton and recent evidence suggests a role for the Formin homology (FH) proteins in these processes. The protein encoded by this gene contains FH domains and belongs to a novel FH protein subfamily implicated in cell polarity. Wnt/Fz signaling activates the small GTPase Rho, a key regulator of cytoskeleton architecture, to control cell polarity and movement during development. Activation requires Dvl-Rho complex formation, an assembly mediated by this gene product, which is thought to function as a scaffolding protein. Evidence of alternative splicing has been observed for this gene but the full-length nature of these variants has not been determined.

Sequence

Synthetic peptide located within the following region: GNTVQYWLLLDRIIQQIVIQNDKGQDPDSTPLENFNIKNVVRMLVNENEV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuangshuang Zhao et al.
Cytoskeleton (Hoboken, N.J.), 77(1-2), 16-24 (2019-12-11)
Formins and tropomyosins (Tpms) are two central components of the microfilaments. Formins are involved in the nucleation and polymerization of actin filaments, and Tpms form along the actin stress fibers to regulate their dynamics. However, the correlation between formins and
Massimo Venditti et al.
Biomolecules, 10(5) (2020-05-02)
Cell differentiation during spermatogenesis requires a proper actin dynamic, regulated by several proteins, including formins. Disheveled-Associated-Activator of Morphogenesis1 (DAAM1) belongs to the formins and promotes actin polymerization. Our results showed that oral D-Aspartate (D-Asp) administration, an excitatory amino acid, increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service