EPM1324-9910
Slide-Out Platform for Eppendorf® New Brunswick™ I26/I26R Incubator Shaker
for 2.8 L Fernbach flasks, dedicated, non-Interchangeable, aluminum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
aluminum
feature
dedicated
for 2.8 L Fernbach flasks
non-Interchangeable
manufacturer/tradename
Eppendorf® M1324-9910 (NA)
size
76 cm × 46 cm
capacity
(6 x 2.8 L Fernback flasks)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Slide-Out platform for Eppendorf New Brunswick I26 and I26 R incubated shaker.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
New Brunswick is a trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service