EPK0641-1690
Eppendorf® Chest Freezer Rack
Holds 13 x 53 mm/ 2 in boxes, (1 x 13) configuration, aluminum, side access
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
aluminum
feature
(1 x 13) configuration
Holds 13 x 53 mm/ 2 in boxes
max. box size [mm]: 133 x 133
side access
manufacturer/tradename
Eppendorf® K0641-1690 (Global)
technique(s)
cryopreservation: suitable
D × W × H
136 mm (5.4 in.) × 139 mm (5.5 in.) × 751 mm (29.6 in.)
Looking for similar products? Visit Product Comparison Guide
General description
for ULT chest freezer: Innova C585 (max. 416 boxes), Innova C760 (max. 572 boxes), CryoCube FC660/ FC660h (max. 520 boxes)max. box number per rack: 13 (1 x 13)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service