EP5943000131
Eppendorf® Centrifuge 5910 Ri G
refrigerated, without rotor, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Synonym(s):
Eppendorf® Centrifuge 5910 Ri G
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
40161701
Recommended Products
feature
CE compliant
CSA approved
refrigerated
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5943000131
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Centrifuge 5910 Ri G
- refrigerated
- with touchscreen interface
- Big Volume Harvesting Solution
- with rotor S-4xUniversal incl. universal buckets
- 1000 ml bottles and 1000 mL adapters
- 120V, US
The new refrigerated benchtop Centrifuge 5910 Ri stands out for its ease of use and versatility: The large 7-inch VisioNize touch interface with an intuitive user experience provides fast and error-free operation, ensuring efficient and reproducible runs. Additionally, the documentation function and unique user management support your quality control with multiple authorization levels, password protection, and documentation of all runs and user activities. Together with a comprehensive portfolio of fixed-angle and swing-bucket rotors, this centrifuge accommodates a particularly broad spectrum of applications, making it the perfect choice for multiuser labs with diverse requirements.
Application
Centrifugation
Features and Benefits
Super versatile multipurpose centrifuge with high capacity for broad range of consumables, refrigerated for temperature sensitive samples
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service