Skip to Content
Merck
All Photos(1)

Key Documents

Sp2/0-Ag14-TurboDoma

3092501, mouse unknown/unspecified, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Sp2/0-Ag14-TurboDoma, 03092501

biological source

mouse unknown/unspecified

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Mouse hybridoma, non-producing, serum-free

Cell Line Description

Sp2/0-Ag14 cell line (Sigma Catalogue number. 85072401) adapted to grow in the chemically defined protein- and peptide-free medium TurboDoma (Cell Culture Technologies). Suitable for use as a fusion parent for serum-free generation of hybridomas as well as a host for the production of recombinant proteins.

Application

Fusion partner for generation of serum-free hybridomas, host for the expression of recombinant genes.

Culture Medium

TurboDoma medium + 4mM Glutamine. When freezing cells down use TurboDoma medium (50:50 conditioned medium:fresh medium) + 4mM Glutamine + 10% DMSO.

Subculture Routine

Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Transfer the contents to a centrifuge tube and slowly add 5-10ml of pre-warmed growth media. Remove a sample for counting. Centrifuge at 100g for 2-3 minutes to pellet cells and seed at a relatively high density of 5-7 x 105 cells/ml. Leave culture flask upright and observe regularly until viable proliferating cells are seen. Once established use a split ratio of 1:2 approximately every 4 to 5 days; 5% CO2; 37°C..

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

TurboDoma is a trademark of Cell Culture Technologies

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service