Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
technique(s)
X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
SRM 1276A_cert
SRM 1276A_SDS
Application
Other Notes
Copper (Cu), Nickel (Ni), Iron (Fe), Zinc (Zn), Lead (Pb), Manganese (Mn), Antimony (Sb), Tin (Sn), Phosphorus (P), Cadmium (Cd), Selenium (Se), Magnesium (Mg), Cobalt (Co)
Reference/Informational value is provided for the following analytes.
Silver (Ag), Arsenic (As), Boron (B), Bismuth (Bi), Chromium (Cr), Sulfur (S), Silicon (Si), Tellurium (Te), Titanium (Ti)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 1 - Pyr. Sol. 1 - Skin Sens. 1 - STOT RE 1 Inhalation
Target Organs
Lungs
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service