Skip to Content
Merck
All Photos(1)

Key Documents

5.31066

Sigma-Aldrich

Remodelin

Synonym(s):

Remodelin, Cyclopentylidene-(4-(4-cyanophenyl)thiazol-2-yl)hydrazine, HBr, 4-(4-Chlorophenyl)-2-(2-cyclopentylidenehydrazinyl)thiazole, HBr, 4-(2-(2-Cyclopentylidenehydrazino)-1,3-thiazol-4-yl)benzonitrile, HBr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4S · xHBr
CAS Number:
Molecular Weight:
282.36 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥94% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

tan

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

C1CCC(=NNC2=NC(=CS2)C3=CC=C(C=C3)C#N)C1.Br

General description

A cell-permeable, potent, and stable analog of CPTH2 (Cat. No. 382111) that restores nuclear shape and chromatin compactin in AG11498 and AG06297 progeric fibroblasts from Hutchinson-Gilford progeria syndrome (HGPS) patients and in lamin A/C-depleted U2OS cells by inhibiting the activity of N-acetyltransferase 10 (NAT10; 10-50 µM). However, it does not affect non-laminopathic Werner syndrome cells. Improves DNA replication and enhances cell proliferation capacity. Also shown to prevent farnesyltransferase inhibitor-induced nuclear shape defects in normal fibroblasts and in U2OS cells and improves nuclear morphology in siLMNA (lamin A/C depletion by small interfering RNA transfection) treated cells. Does not affect Golgi apparatus integrity or tubulin polymer assembly and its effects are independent of mitosis.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NAT10
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Larrieu, D., et al. 2014. Science344, 527.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Larrieu et al.
Science (New York, N.Y.), 344(6183), 527-532 (2014-05-03)
Down-regulation and mutations of the nuclear-architecture proteins lamin A and C cause misshapen nuclei and altered chromatin organization associated with cancer and laminopathies, including the premature-aging disease Hutchinson-Gilford progeria syndrome (HGPS). Here, we identified the small molecule "Remodelin" that improved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service