17-10398
In Vitro Vascular Permeability Imaging Assay (Green)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84
Recommended Products
species reactivity
all
Quality Level
technique(s)
cell based assay: suitable
detection method
fluorometric
Application
The EMD Millipore In Vitro Vascular Permeability Imaging Assay kit is ideal for interrogating compounds that may disrupt or protect an endothelial monolayer. Each In Vitro Vascular Permeability Imaging Assay kit contains sufficient reagents for the coating of four 8-well glass chamber slides, although alternate substrate formats may also be utilized. The EMD Millipore In Vitro Vascular Permeability Imaging Assay is intended for research use only, not for diagnostic or therapeutic applications.
Components
Poly-L-Lysine, 2X DAPI Glutaraldehyde, 16X Biotin-Gelatin Fluorescein-Streptavidin VE Cadherin Primary Antibody Donkey anti-Mouse IgG, Cy3™ Conjugate Secondary
Storage and Stability
Store Poly-L-Lysine and DAPI at 2-8°C. Store Biotin-Gelatin, Glutaraldehyde, fluorescein-streptavidin, anti-VE cadherin, and Donkey anti-Mouse IgG, Cy3™ conjugate at -20°C.Use within 4 months from date of receipt.
Legal Information
Cy3 is a trademark of Cytiva
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service