Skip to Content
Merck
All Photos(1)

Key Documents

1.46371

Millipore

Buffered NaCl-Peptone Solution + 0.1% Tween® 80

bottle capacity 275 mL, bottle filling volume 90 mL, closure type, White screw cap for wide neck, pack of 6 bottles

Synonym(s):

Buffered Saline Peptone Water with 0.1% Tween®, Buffered Sodium Chloride Peptone Solution with 0.1% Polysorbate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.13
JP 5.02
USP 62

Quality Level

description

For the microbial examination of non-sterile pharmaceutical products

form

liquid

shelf life

12 mo.

feature

closure type White screw cap for wide neck
ready-to-use

packaging

pack of 6 bottles

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

275 mL

bottle filling volume

90 mL

pH

7.0

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

components

tween: 0.1%

storage temp.

2-25°C

General description

Buffered NaCl-Peptone Solution with 0.1% Tween® 80 has a low content of peptone along with phosphate buffer and sodium chloride that maintain the viability of microorganisms present without supporting growth. Tween® 80 assists the suspension of poorly wettable substances and also serve as a neutralizing agent for disinfectants, such as quaternary ammonium compounds, iodines, and parabens.

Application

Buffered NaCl-Peptone Solution with 0.1% Tween® 80 is designed for the microbial examination of non-sterile pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Our media are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service