MAS-100 ISO NT® Elbow Joint with 2 Tri-Clamps is an additional accessory designed to be specifically used for MAS-100 ISO NT® and MAS-100 ISO MH®. The MAS 100 Iso line microbial air samplers have been specially developed for monitoring and measuring the microbiological contamination at fixed sampling locations in cleanrooms, RABS and isolators. The two tri-clamp of 21.7mm internal diameter and 50.5 mm external diameter is used to connect the stainless steel or reinforced silicone tubing with the sampling head base and the instrument. The elbow joints aid in connecting at a 90° angle bend wherever necessary.
Application
MAS-100 ISO NT® Elbow Joint with 2 Tri-Clamps is an additional accessory used in connections of the instrument. MAS-100 Iso family devices are extensively used in Pharmaceutical applications.
Legal Information
MAS-100 ISO is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.