BR780773
BRAND® screw caps with tamper evident screw cap for micro tubes
purple
Synonym(s):
Accessories, Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
polypropylene
purple
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 780773
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Applications: -196 °C to +121 °C.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service