BR37489
BRAND® volumetric flask BLAUBRAND® USP
capacity 250 mL, joint: 14/23, polypropylene stopper
Synonym(s):
BLAUBRAND®, USP, Volumetric flasks
About This Item
material
amber
borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint ST/NS neck14/23
joint: 14/23
feature
class A DE-M marking. To DIN EN ISO 1042. Calibrated to contain (TC, In). With USP batch certificate.
DE-M marking
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37489
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service