BR114516
BRAND® test tube, graduated, glass
capacity 20 mL, with PP-stopper NS 14/23
Synonym(s):
Test tubes, Tubes
About This Item
Recommended Products
material
borosilicate glass tube
cap (taper)
colorless tube
polypropylene cap
round bottom
white cap
sterility
non-sterile
feature
graduations
neck thread Push fit
with PP-stopper NS 14/23
packaging
pack of 10 ea
manufacturer/tradename
BRAND 114516
capacity
20 mL
diam. × L
17 mm × 200 mm
tube diam.
17 mm
fitting
(NS 14/23)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- BRAND® graduated test tubes are versatile to fit most laboratory applications.
- Manufactured from clear borosilicate glass for easy viewing of contents.
- Includes a polypropylene cap for optimal sample recovery.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service