Skip to Content
Merck
All Photos(1)

Documents

802840

Sigma-Aldrich

Bis(2-ethylhexyl)-d34 phosphate

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D34HO4P
Molecular Weight:
356.63
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

mass shift

M+34

SMILES string

O=P(OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C16H35O4P/c1-5-9-11-15(7-3)13-19-21(17,18)20-14-16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3,(H,17,18)/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D,16D

InChI key

SEGLCEQVOFDUPX-RFETUZSTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service