617229
Dimethyl [2-oxo-2-(cyclohexyl-d11)ethyl]phosphonate
98 atom % D
Synonym(s):
Dimethyl (2-cyclohexyl-d11-2-oxoethyl)phosphonate
About This Item
Recommended Products
isotopic purity
98 atom % D
reaction suitability
reaction type: C-C Bond Formation
mass shift
M+11
SMILES string
[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(=O)CP(=O)(OC)OC)C([2H])([2H])C1([2H])[2H]
InChI
1S/C10H19O4P/c1-13-15(12,14-2)8-10(11)9-6-4-3-5-7-9/h9H,3-8H2,1-2H3/i3D2,4D2,5D2,6D2,7D2,9D
InChI key
FTZNMXSYERMMBC-QFAYMVOLSA-N
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service