- Mapping the substrate-binding site of a human class mu glutathione transferase using nuclear magnetic resonance spectroscopy.
Mapping the substrate-binding site of a human class mu glutathione transferase using nuclear magnetic resonance spectroscopy.
The substrate-binding site of a human muscle class mu glutathione transferase has been characterized using high-resolution nuclear magnetic resonance spectroscopy. Isotopic labeling has been used to simplify one-dimensional proton NMR spectra of the Tyr and His residues in the enzyme and two-dimensional carbon-proton spectra of the Ala and Met residues in the enzyme. The resonance lines from 8 of the 12 Tyr residues have been assigned using site-directed mutagenesis. Replacement of Tyr7 with Phe reduced the activity of the enzyme 100-fold. The proximity of His, Tyr, Ala, and Met residues to the active site has been determined using a nitroxide-labeled substrate analogue. This substrate analogue binds with high affinity (Keq = 10(6) M-1) to the enzyme and is a competitive inhibitor. None of the His residues are within 17 A of the active site. Three of the assigned Tyr residues are greater than 17 A from the active site. Quantitative measurement of paramagnetic line broadening of five additional Tyr residues places them within 13-17 A from the active site. Broadening of the Ala and Met resonance lines by the spin-labeled substrate indicates that three Ala residues are 9-16 A from the nitroxide, three Met residues are less than 9 A from the nitroxide, and two Met residues are 9-16 A from the nitroxide.(ABSTRACT TRUNCATED AT 250 WORDS)