
Sulfur in di-n-butyl sulfide

Synonym(s):
Dibutyl sulfide, 1,1′-Thiobisbutane, Butyl sulfide, NSC 8460
Linear Formula:
CH3(CH2)3S(CH2)3CH3
CAS Number:
Molecular Weight:
146.29
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1